Tetrabutylammonium bromide, 99+%
Tetrabutylammonium bromide, 99+%
Tetrabutylammonium bromide, 99+%
Tetrabutylammonium bromide, 99+%
Thermo Scientific Chemicals

Tetrabutylammonium bromide, 99+%

CAS: 1643-19-2 | C16H36BrN | 322.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
50 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 185680250
Price (USD)
51.65
Online Exclusive
56.80
Save 5.15 (9%)
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
51.65
Online Exclusive
56.80
Save 5.15 (9%)
Each
Add to cart
Chemical Identifiers
CAS1643-19-2
IUPAC Nametetrabutylazanium bromide
Molecular FormulaC16H36BrN
InChI KeyJRMUNVKIHCOMHV-UHFFFAOYSA-M
SMILES[Br-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Infrared spectrumConforms
Titration Argentometric>=99.0 %
Water=<1 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link