Tetrabutylammonium bromide, 99+%, Thermo Scientific Chemicals
Tetrabutylammonium bromide, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrabutylammonium bromide, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
50 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 185681000
Price (USD)/ Each
75.65
Online exclusive
84.00 
Save 8.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
75.65
Online exclusive
84.00 
Save 8.35 (10%)
Add to cart
Tetrabutylammonium bromide, 99+%, Thermo Scientific Chemicals
Catalog number185681000
Price (USD)/ Each
75.65
Online exclusive
84.00 
Save 8.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1643-19-2
IUPAC Nametetrabutylazanium bromide
Molecular FormulaC16H36BrN
InChI KeyJRMUNVKIHCOMHV-UHFFFAOYSA-M
SMILES[Br-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration Argentometric>=99.0 %
Water=<1 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only