1,4-Benzodioxan-6-amine, 99%, Thermo Scientific Chemicals
1,4-Benzodioxan-6-amine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Benzodioxan-6-amine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
50 g
Packaging:
Glass bottle
Catalog number 185782500
Price (USD)/ Each
641.00
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
641.00
Add to cart
1,4-Benzodioxan-6-amine, 99%, Thermo Scientific Chemicals
Catalog number185782500
Price (USD)/ Each
641.00
-
Add to cart
Chemical Identifiers
CAS22013-33-8
IUPAC Name2,3-dihydro-1,4-benzodioxin-6-amine
Molecular FormulaC8H9NO2
InChI KeyBZKOZYWGZKRTIB-UHFFFAOYSA-N
SMILESNC1=CC=C2OCCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5970 to 1.6010 (20°C, 589 nm)
Appearance (Color)Yellow to dark brown
Appearance (Form)Solid or liquid
Infrared spectrumConforms
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only