Thermo Scientific Chemicals

2-Chloropyrimidine, 98+%, Thermo Scientific Chemicals

Catalog number: 185840100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Chloropyrimidine, 98+%, Thermo Scientific Chemicals

Catalog number: 185840100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 185840100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1722-12-9
IUPAC Name
2-chloropyrimidine
Molecular Formula
C4H3ClN2
InChI Key
UNCQVRBWJWWJBF-UHFFFAOYSA-N
SMILES
ClC1=NC=CC=N1
Appearance (Color)
Yellow to light orange
Appearance (Form)
Crystalline powder or crystals
Melting point
62°C to 66°C
GC
>=98.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text