1,4-Diiodobenzene, 98%, Thermo Scientific Chemicals
1,4-Diiodobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Diiodobenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 185970500
Price (USD)/ Each
92.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
109.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
92.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
109.00 
Save 16.35 (15%)
Add to cart
1,4-Diiodobenzene, 98%, Thermo Scientific Chemicals
Catalog number185970500
Price (USD)/ Each
92.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
109.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
CAS624-38-4
IUPAC Name1,4-diiodobenzene
Molecular FormulaC6H4I2
InChI KeyLFMWZTSOMGDDJU-UHFFFAOYSA-N
SMILESIC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to tan
Infrared spectrumConforms
Melting point128°C to 133°C
GC>=97.5 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only