Thermo Scientific Chemicals

MEM chloride, 94%, Thermo Scientific Chemicals

Catalog number: 185980250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

MEM chloride, 94%, Thermo Scientific Chemicals

Catalog number: 185980250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 185980250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3970-21-6
IUPAC Name
1-(chloromethoxy)-2-methoxyethane
Molecular Formula
C4H9ClO2
InChI Key
BIAAQBNMRITRDV-UHFFFAOYSA-N
SMILES
COCCOCCl
Refractive index
1.4260 to 1.4280 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=92 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text