2,2-Dibromopropane, 98%, Thermo Scientific Chemicals
2,2-Dibromopropane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2-Dibromopropane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Packaging:
Glass bottle
Catalog number 186060050
Price (USD)/ Each
77.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
90.60 
Save 12.95 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
77.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
90.60 
Save 12.95 (14%)
Add to cart
2,2-Dibromopropane, 98%, Thermo Scientific Chemicals
Catalog number186060050
Price (USD)/ Each
77.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
90.60 
Save 12.95 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name2,2-dibromopropane
Molecular FormulaC3H6Br2
InChI KeyARITXYXYCOZKMU-UHFFFAOYSA-N
SMILESCC(C)(Br)Br
Molecular Weight (g/mol)201.89
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.4950 to 1.4990 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only