Thermo Scientific Chemicals

1-Indanol, 98%, Thermo Scientific Chemicals

Catalog number: 186110500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Indanol, 98%, Thermo Scientific Chemicals

Catalog number: 186110500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 186110500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6351-10-6
IUPAC Name
2,3-dihydro-1H-inden-1-ol
Molecular Formula
C9H10O
InChI Key
YIAPLDFPUUJILH-UHFFFAOYNA-N
SMILES
OC1CCC2=CC=CC=C12
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals and/or chunks or fused solid
Melting point
50°C to 55°C
Appearance (Color)
White to yellow to beige to light brown
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text