Thermo Scientific Chemicals

1-Methylindole, 98+%, Thermo Scientific Chemicals

Catalog number: 186300250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Methylindole, 98+%, Thermo Scientific Chemicals

Catalog number: 186300250
25 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 186300250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
603-76-9
IUPAC Name
1-methyl-1H-indole
Molecular Formula
C9H9N
InChI Key
BLRHMMGNCXNXJL-UHFFFAOYSA-N
SMILES
CN1C=CC2=CC=CC=C12
Refractive index
1.6050 to 1.6080 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text