Benzimidazole, 98%
Benzimidazole, 98%
Benzimidazole, 98%
Thermo Scientific Chemicals

Benzimidazole, 98%

CAS: 51-17-2 | C7H6N2 | 118.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 186421000
Price (USD)
53.65
Online Exclusive
59.20
Save 5.55 (9%)
Each
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.20
Save 5.55 (9%)
Each
Add to cart
Chemical Identifiers
CAS51-17-2
IUPAC Name1H-1,3-benzodiazole
Molecular FormulaC7H6N2
InChI KeyHYZJCKYKOHLVJF-UHFFFAOYSA-N
SMILESN1C=NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to brown
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point171°C to 174°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link