Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number 186430010
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
Add to cart
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Catalog number186430010
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS144-62-7
IUPAC Nameoxalic acid
Molecular FormulaC2H2O4
InChI KeyMUBZPKHOEPUJKR-UHFFFAOYSA-N
SMILESOC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with NaOH97.5 to 102.5 %
Appearance (Color)White to almost white
Infrared spectrumConforms
Melting point189°C to 191°C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only