Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 186430050
Price (USD)/ Each
455.65
Online exclusive
536.00 
Save 80.35 (15%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
455.65
Online exclusive
536.00 
Save 80.35 (15%)
Add to cart
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Catalog number186430050
Price (USD)/ Each
455.65
Online exclusive
536.00 
Save 80.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameoxalic acid
Molecular FormulaC2H2O4
InChI KeyMUBZPKHOEPUJKR-UHFFFAOYSA-N
SMILESOC(=O)C(O)=O
Molecular Weight (g/mol)90.03
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with NaOH97.5 to 102.5 %
Appearance (Color)White to almost white
Infrared spectrumConforms
Melting point189°C to 191°C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only