Oxalic acid, 98%, anhydrous
Oxalic acid, 98%, anhydrous
Oxalic acid, 98%, anhydrous
Oxalic acid, 98%, anhydrous
Oxalic acid, 98%, anhydrous
Thermo Scientific Chemicals

Oxalic acid, 98%, anhydrous

CAS: 144-62-7 | C2H2O4 | 90.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 186432500
Price (USD)
44.65
Special offer
Online exclusive
Ends: 26-Sep-2025
54.80
Save 10.15 (19%)
Each
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 26-Sep-2025
54.80
Save 10.15 (19%)
Each
Add to cart
Chemical Identifiers
CAS144-62-7
IUPAC Nameoxalic acid
Molecular FormulaC2H2O4
InChI KeyMUBZPKHOEPUJKR-UHFFFAOYSA-N
SMILESOC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with NaOH97.5 to 102.5 %
Appearance (Color)White to almost white
Infrared spectrumConforms
Melting point189°C to 191°C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Share catalog number, name or link