Thermo Scientific Chemicals

2-Chlorobenzyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: 186532500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

2-Chlorobenzyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: 186532500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 186532500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
611-19-8
IUPAC Name
1-chloro-2-(chloromethyl)benzene
Molecular Formula
C7H6Cl2
InChI Key
BASMANVIUSSIIM-UHFFFAOYSA-N
SMILES
ClCC1=CC=CC=C1Cl
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to light yellow
GC
>=98.0 %
Refractive index
1.5580 to 1.5600 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text