Thermo Scientific Chemicals

Bromotrimethylsilane, 98%, Thermo Scientific Chemicals

Catalog number: 186655000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Bromotrimethylsilane, 98%, Thermo Scientific Chemicals

Catalog number: 186655000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 186655000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2857-97-8
Titration Argentometric
>=97.5 %
Appearance (Color)
Clear colorless to yellow
Refractive index
1.4200 to 1.4250 (20°C, 589 nm)
Infrared spectrum
Conforms
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Silylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text