Methoxyacetic acid, 97+%, Thermo Scientific Chemicals
Methoxyacetic acid, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methoxyacetic acid, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
25 mL
1 L
Packaging:
Glass bottle
Catalog number 186772500
Price (USD)/ Each
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS625-45-6
IUPAC Name2-methoxyacetic acid
Molecular FormulaC3H6O3
InChI KeyRMIODHQZRUFFFF-UHFFFAOYSA-N
SMILESCOCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.4150 to 1.4180 (20°C, 589 nm)
Appearance (Form)Liquid
Titration with NaOH97.0 to 103.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only