12-Crown-4, 97%, Thermo Scientific Chemicals
12-Crown-4, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

12-Crown-4, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
2.5 g
5 g
Packaging:
Glass bottle
Catalog number 187020250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS294-93-9
IUPAC Name1,4,7,10-tetraoxacyclododecane
Molecular FormulaC8H16O4
InChI KeyXQQZRZQVBFHBHL-UHFFFAOYSA-N
SMILESC1COCCOCCOCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Liquid after melting
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4600 to 1.4630 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only