Thermo Scientific Chemicals

1-Bromotetradecane, 98%, Thermo Scientific Chemicals

Catalog number: 187381000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Bromotetradecane, 98%, Thermo Scientific Chemicals

Catalog number: 187381000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 187381000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-71-0
IUPAC Name
1-bromotetradecane
Molecular Formula
C14H29Br
InChI Key
KOFZTCSTGIWCQG-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCBr
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Form)
Liquid
Refractive index
1.4580 to 1.4620 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text