Thermo Scientific Chemicals

Iodotrimethylsilane, 95-97%, stabilized, Thermo Scientific Chemicals

Catalog number: 187491000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Iodotrimethylsilane, 95-97%, stabilized, Thermo Scientific Chemicals

Catalog number: 187491000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 187491000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16029-98-4
IUPAC Name
iodotrimethylsilane
Molecular Formula
C3H9ISi
InChI Key
CSRZQMIRAZTJOY-UHFFFAOYSA-N
SMILES
C[Si](C)(C)I
Appearance (Color)
Clear colorless to red
Infrared spectrum
Conforms
Titration Argentometric
>=95 % (Ex I)
Appearance (Form)
Liquid
Refractive index
1.4700 to 1.4800 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text