Iodotrimethylsilane, 95-97%, stabilized, Thermo Scientific Chemicals
Iodotrimethylsilane, 95-97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iodotrimethylsilane, 95-97%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
25 mL
100 mL
Packaging:
View all
Glass Bottle
Catalog number 187492500
Price (USD)/ Each
-
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nameiodotrimethylsilane
Molecular FormulaC3H9ISi
InChI KeyCSRZQMIRAZTJOY-UHFFFAOYSA-N
SMILESC[Si](C)(C)I
Molecular Weight (g/mol)200.09
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to red
Infrared spectrumConforms
Titration Argentometric>=95 % (Ex I)
Appearance (Form)Liquid
Refractive index1.4700 to 1.4800 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only