Thermo Scientific Chemicals

tert-Butylchlorodiphenylsilane, 98%, Thermo Scientific Chemicals

Catalog number: 187500100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

tert-Butylchlorodiphenylsilane, 98%, Thermo Scientific Chemicals

Catalog number: 187500100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 187500100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58479-61-1
IUPAC Name
tert-butyl(chloro)diphenylsilane
Molecular Formula
C16H19ClSi
InChI Key
MHYGQXWCZAYSLJ-UHFFFAOYSA-N
SMILES
CC(C)(C)[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance
May darken on storage
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.5665 to 1.5685 (20°C, 589 nm)
Appearance (Color)
Clear to hazy colorless to yellow to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text