2-Nitroimidazole, 98%, Thermo Scientific Chemicals
2-Nitroimidazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Nitroimidazole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Packaging:
Glass bottle
Catalog number 187570010
Price (USD)/ Each
401.00
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
401.00
Add to cart
2-Nitroimidazole, 98%, Thermo Scientific Chemicals
Catalog number187570010
Price (USD)/ Each
401.00
-
Add to cart
Chemical Identifiers
CAS527-73-1
IUPAC Name2-nitro-1H-imidazole
Molecular FormulaC3H3N3O2
InChI KeyYZEUHQHUFTYLPH-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=NC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to khaki
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only