cis-1,4-Dichloro-2-butene, 95%
cis-1,4-Dichloro-2-butene, 95%
cis-1,4-Dichloro-2-butene, 95%
cis-1,4-Dichloro-2-butene, 95%
Thermo Scientific Chemicals

cis-1,4-Dichloro-2-butene, 95%

CAS: 1476-11-5 | C4H6Cl2 | 125 g/mol
Have Questions?
Catalog NumberQuantityPackaging
18760025025 mLGlass bottle
Catalog number 187600250
Price (USD)
116.65
Special offer
Online exclusive
Ends: 26-Sep-2025
144.00
Save 27.35 (19%)
Each
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
116.65
Special offer
Online exclusive
Ends: 26-Sep-2025
144.00
Save 27.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS1476-11-5
IUPAC Name(2Z)-1,4-dichlorobut-2-ene
Molecular FormulaC4H6Cl2
InChI KeyFQDIANVAWVHZIR-UPHRSURJSA-N
SMILESClC\C=C/CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4884 to 1.4914 (20°C, 589 nm)
GC>=94 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link