Thermo Scientific Chemicals

Iron(0) pentacarbonyl, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 187621000
Each
Thermo Scientific Chemicals

Iron(0) pentacarbonyl, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 187621000
Each
Catalog number: 187621000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13463-40-6
IUPAC Name
pentakis(methanidylidyneoxidanium) iron
Molecular Formula
C5FeO5
InChI Key
FYOFOKCECDGJBF-UHFFFAOYSA-N
SMILES
[Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Additional info
Material may have to be filtered through glass
Additional info
wool before use. Keep residue moist,
Additional info
may be pyrophoric if allowed to dry.
Appearance (Color)
Yellow-orange to brown
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text