2-Thiopheneacetic acid, 98%, Thermo Scientific Chemicals
2-Thiopheneacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Thiopheneacetic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 187730250
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
69.10 
Save 10.45 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
69.10 
Save 10.45 (15%)
Add to cart
2-Thiopheneacetic acid, 98%, Thermo Scientific Chemicals
Catalog number187730250
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
69.10 
Save 10.45 (15%)
-
Add to cart
Chemical Identifiers
CAS1918-77-0
IUPAC Name2-(thiophen-2-yl)acetic acid
Molecular FormulaC6H6O2S
InChI KeySMJRBWINMFUUDS-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point61°C to 66°C
Titration with NaOH97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only