4-Isopropylbenzyl alcohol, 97%, Thermo Scientific Chemicals
4-Isopropylbenzyl alcohol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Isopropylbenzyl alcohol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 187801000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name[4-(propan-2-yl)phenyl]methanol
Molecular FormulaC10H14O
InChI KeyOIGWAXDAPKFNCQ-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(CO)C=C1
Molecular Weight (g/mol)150.22
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.5180 to 1.5210 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only