Thermo Scientific Chemicals

4-Isopropylbenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog number: 187805000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Isopropylbenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog number: 187805000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 187805000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
536-60-7
IUPAC Name
[4-(propan-2-yl)phenyl]methanol
Molecular Formula
C10H14O
InChI Key
OIGWAXDAPKFNCQ-UHFFFAOYSA-N
SMILES
CC(C)C1=CC=C(CO)C=C1
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.5180 to 1.5210 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text