4-Chloro-2-nitrobenzyl chloride, 98%, Thermo Scientific Chemicals
4-Chloro-2-nitrobenzyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloro-2-nitrobenzyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 187950010
Price (USD)/ Each
51.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
51.00
Add to cart
4-Chloro-2-nitrobenzyl chloride, 98%, Thermo Scientific Chemicals
Catalog number187950010
Price (USD)/ Each
51.00
-
Add to cart
Chemical Identifiers
CAS938-71-6
IUPAC Name1-chloro-3-(chloromethyl)-5-nitrobenzene
Molecular FormulaC7H5Cl2NO2
InChI KeyQBIQAXWMONJLNW-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(Cl)=CC(CCl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting solid
NMR (1H-NMR)Consistent with structure
Appearance (Color)Light yellow to tan
Infrared spectrumConforms
Melting point39°C to 44°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only