Thermo Scientific Chemicals

4-Chloro-2-nitrobenzyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 187950050
5 g, Each
Thermo Scientific Chemicals

4-Chloro-2-nitrobenzyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 187950050
5 g, Each
Quantity
Catalog number: 187950050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
938-71-6
IUPAC Name
1-chloro-3-(chloromethyl)-5-nitrobenzene
Molecular Formula
C7H5Cl2NO2
InChI Key
QBIQAXWMONJLNW-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC(Cl)=CC(CCl)=C1
Appearance (Form)
Low melting solid
NMR (1H-NMR)
Consistent with structure
Appearance (Color)
Light yellow to tan
Infrared spectrum
Conforms
Melting point
39°C to 44°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text