Thiomorpholine, 97%, Thermo Scientific Chemicals
Thiomorpholine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thiomorpholine, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 187970250
Price (USD)/ Each
136.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
161.00 
Save 24.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
136.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
161.00 
Save 24.35 (15%)
Add to cart
Thiomorpholine, 97%, Thermo Scientific Chemicals
Catalog number187970250
Price (USD)/ Each
136.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
161.00 
Save 24.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namehydrogen thiomorpholine chloride
Molecular FormulaC4H10ClNS
InChI KeyQSJAHJYXDRUZMY-UHFFFAOYSA-N
SMILES[H+].[Cl-].C1CSCCN1
Molecular Weight (g/mol)139.64
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5370 to 1.5390 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only