3-Furanmethanol, 99%, Thermo Scientific Chemicals
3-Furanmethanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Furanmethanol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 188080050
Price (USD)/ Each
187.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
187.00
Add to cart
3-Furanmethanol, 99%, Thermo Scientific Chemicals
Catalog number188080050
Price (USD)/ Each
187.00
-
Add to cart
Chemical Identifiers
CAS4412-91-3
IUPAC Name(furan-3-yl)methanol
Molecular FormulaC5H6O2
InChI KeySTJIISDMSMJQQK-UHFFFAOYSA-N
SMILESOCC1=COC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4835 to 1.4855 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only