Thermo Scientific Chemicals

4-Biphenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 188150100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Biphenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 188150100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 188150100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5728-52-9
IUPAC Name
2-{[1,1'-biphenyl]-4-yl}acetic acid
Molecular Formula
C14H12O2
InChI Key
QRZAKQDHEVVFRX-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=C(C=C1)C1=CC=CC=C1
Appearance (Color)
White to beige
Melting point
158°C to 165°C
Infrared spectrum
Conforms
Appearance (Form)
Powder
Titration with TBAH
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text