4-Biphenylacetic acid, 98%, Thermo Scientific Chemicals
4-Biphenylacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Biphenylacetic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number 188150250
Price (USD)/ Each
110.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
110.00
Add to cart
4-Biphenylacetic acid, 98%, Thermo Scientific Chemicals
Catalog number188150250
Price (USD)/ Each
110.00
-
Add to cart
Chemical Identifiers
CAS5728-52-9
IUPAC Name2-{[1,1'-biphenyl]-4-yl}acetic acid
Molecular FormulaC14H12O2
InChI KeyQRZAKQDHEVVFRX-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Melting point158°C to 165°C
Infrared spectrumConforms
Appearance (Form)Powder
Titration with TBAH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only