Thermo Scientific Chemicals

1,3-Benzenedimethanol, 98%, Thermo Scientific Chemicals

Catalog number: 188200100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

1,3-Benzenedimethanol, 98%, Thermo Scientific Chemicals

Catalog number: 188200100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 188200100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
626-18-6
IUPAC Name
[3-(hydroxymethyl)phenyl]methanol
Molecular Formula
C8H10O2
InChI Key
YWMLORGQOFONNT-UHFFFAOYSA-N
SMILES
OCC1=CC(CO)=CC=C1
Appearance (Color)
White to beige
Appearance (Form)
Powder and/or chunks
Infrared spectrum
Conforms
Melting point
54°C to 59°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text