3-Fluoropyridine, 99%, Thermo Scientific Chemicals
3-Fluoropyridine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluoropyridine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 188300050
Price (USD)/ Each
156.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
156.00
Add to cart
3-Fluoropyridine, 99%, Thermo Scientific Chemicals
Catalog number188300050
Price (USD)/ Each
156.00
-
Add to cart
Chemical Identifiers
IUPAC Name3-fluoropyridine
Molecular FormulaC5H4FN
InChI KeyCELKOWQJPVJKIL-UHFFFAOYSA-N
SMILESFC1=CN=CC=C1
Molecular Weight (g/mol)97.09
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless to yellow
Refractive index1.4720 to 1.4750 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only