Thermo Scientific Chemicals

2,3,5,6-Tetrafluorophenol, 98%, Thermo Scientific Chemicals

Catalog number: 188390050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2,3,5,6-Tetrafluorophenol, 98%, Thermo Scientific Chemicals

Catalog number: 188390050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 188390050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
769-39-1
IUPAC Name
2,3,5,6-tetrafluorophenol
Molecular Formula
C6H2F4O
InChI Key
PBYIIRLNRCVTMQ-UHFFFAOYSA-N
SMILES
OC1=C(F)C(F)=CC(F)=C1F
Appearance (Color)
White
Melting point
33°C to 38°C
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Form)
Low melting crystalline solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text