Thermo Scientific Chemicals

2,3,4,5,6-Pentafluorostyrene, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 188470050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2,3,4,5,6-Pentafluorostyrene, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 188470050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 188470050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
653-34-9
IUPAC Name
1-ethenyl-2,3,4,5,6-pentafluorobenzene
Molecular Formula
C8H3F5
InChI Key
LVJZCPNIJXVIAT-UHFFFAOYSA-N
SMILES
FC1=C(F)C(F)=C(C=C)C(F)=C1F
Appearance (Form)
Liquid
Refractive index
1.4450 to 1.4470 (20°C, 589 nm)
Stabilizer
0.01 to 0.1 % p-tert-Butylcatechol
Appearance (Color)
Colorless to yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text