2,3,4,5,6-Pentafluorostyrene, 97%, stabilized, Thermo Scientific Chemicals
2,3,4,5,6-Pentafluorostyrene, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3,4,5,6-Pentafluorostyrene, 97%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 188470250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS653-34-9
IUPAC Name1-ethenyl-2,3,4,5,6-pentafluorobenzene
Molecular FormulaC8H3F5
InChI KeyLVJZCPNIJXVIAT-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(C=C)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4450 to 1.4470 (20°C, 589 nm)
Stabilizer0.01 to 0.1 % p-tert-Butylcatechol
Appearance (Color)Colorless to yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only