Thermo Scientific Chemicals

4-Morpholinoaniline, 98+%, Thermo Scientific Chemicals

Catalog number: 188650250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Morpholinoaniline, 98+%, Thermo Scientific Chemicals

Catalog number: 188650250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 188650250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2524-67-6
IUPAC Name
4-(morpholin-4-yl)aniline
Molecular Formula
C10H14N2O
InChI Key
PHNDZBFLOPIMSM-UHFFFAOYSA-N
SMILES
NC1=CC=C(C=C1)N1CCOCC1
Appearance (Color)
Brown to purple
Appearance (Form)
Crystalline powder
Melting point
128°C to 132°C
Titration with HClO4
>=98.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text