Ethyl 1-bromocyclobutanecarboxylate, 96%, Thermo Scientific Chemicals
Ethyl 1-bromocyclobutanecarboxylate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 1-bromocyclobutanecarboxylate, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 188710010
Price (USD)/ Each
41.20
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
41.20
Add to cart
Ethyl 1-bromocyclobutanecarboxylate, 96%, Thermo Scientific Chemicals
Catalog number188710010
Price (USD)/ Each
41.20
-
Add to cart
Chemical Identifiers
CAS35120-18-4
IUPAC Nameethyl 1-bromocyclobutane-1-carboxylate
Molecular FormulaC7H11BrO2
InChI KeyUTVNSHXHFRIXMM-UHFFFAOYSA-N
SMILESCCOC(=O)C1(Br)CCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4700 to 1.4720 (20°C, 589 nm)
GC>=95 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only