Thermo Scientific Chemicals

Triisopropyl borate, 98+%, Thermo Scientific Chemicals

Catalog number: 188740800
80 g, Glass bottle, Each
Thermo Scientific Chemicals

Triisopropyl borate, 98+%, Thermo Scientific Chemicals

Catalog number: 188740800
80 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 188740800
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5419-55-6
IUPAC Name
tris(propan-2-yl) borate
Molecular Formula
C9H21BO3
InChI Key
NHDIQVFFNDKAQU-UHFFFAOYSA-N
SMILES
CC(C)OB(OC(C)C)OC(C)C
Appearance (Color)
Clear colorless
Refractive index
1.3755 to 1.3775 (20°C, 589 nm)
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text