Methyl cyclohexanecarboxylate, 98%, Thermo Scientific Chemicals
Methyl cyclohexanecarboxylate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl cyclohexanecarboxylate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
Packaging:
Glass bottle
Catalog number 188820500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4630-82-4
IUPAC Namemethyl cyclohexanecarboxylate
Molecular FormulaC8H14O2
InChI KeyZQWPRMPSCMSAJU-UHFFFAOYSA-N
SMILESCOC(=O)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4420 to 1.4440 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only