Thermo Scientific Chemicals

5,6,7,8-Tetrahydroquinoline, 98%, Thermo Scientific Chemicals

Catalog number: 188871000
100 g, Each
Thermo Scientific Chemicals

5,6,7,8-Tetrahydroquinoline, 98%, Thermo Scientific Chemicals

Catalog number: 188871000
100 g, Each
Quantity
Catalog number: 188871000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10500-57-9
IUPAC Name
5,6,7,8-tetrahydroquinoline
Molecular Formula
C9H11N
InChI Key
YQDGQEKUTLYWJU-UHFFFAOYSA-N
SMILES
C1CCC2=NC=CC=C2C1
Appearance (Color)
Colorless to light brown
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text