DL-10-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-10-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number 189070025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS5872-08-2
IUPAC Name{7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl}methanesulfonic acid
Molecular FormulaC10H16O4S
InChI KeyMIOPJNTWMNEORI-UHFFFAOYNA-N
SMILESCC1(C)C2CCC1(CS(O)(=O)=O)C(=O)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Powder or granular powder
Infrared spectrumConforms
Titration with NaOH97.5 to 102.5 % (On dried substance)
Loss on drying=<1.5 % (105°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only