Thermo Scientific Chemicals

2-(4-Chlorophenoxy)-2-methylpropionic acid, 99%, Thermo Scientific Chemicals

Catalog number: 189090500
50 g, Plastic bottle, Each
Thermo Scientific Chemicals

2-(4-Chlorophenoxy)-2-methylpropionic acid, 99%, Thermo Scientific Chemicals

Catalog number: 189090500
50 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 189090500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
882-09-7
IUPAC Name
2-(4-chlorophenoxy)-2-methylpropanoic acid
Molecular Formula
C10H11ClO3
InChI Key
TXCGAZHTZHNUAI-UHFFFAOYSA-N
SMILES
CC(C)(OC1=CC=C(Cl)C=C1)C(O)=O
Appearance (Form)
Crystalline powder or crystals
Loss on drying
=<1 %
Appearance (Color)
Light yellow to brown
Melting point
120°C to 123°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text