3,4,5-Trimethoxyphenol, 98.5+%, Thermo Scientific Chemicals
3,4,5-Trimethoxyphenol, 98.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4,5-Trimethoxyphenol, 98.5+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 189140250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS642-71-7
IUPAC Name3,4,5-trimethoxyphenol
Molecular FormulaC9H12O4
InChI KeyVTCDZPUMZAZMSB-UHFFFAOYSA-N
SMILESCOC1=CC(O)=CC(OC)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige to brown
Appearance (Form)Powder or crystals or needles
Infrared spectrumConforms
Melting point143°C to 149°C
GC>=98.50 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only