Thermo Scientific Chemicals

3-(3,4,5-Trimethoxyphenyl)propionic acid, 98%, Thermo Scientific Chemicals

Catalog number: 189160250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3-(3,4,5-Trimethoxyphenyl)propionic acid, 98%, Thermo Scientific Chemicals

Catalog number: 189160250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 189160250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25173-72-2
IUPAC Name
3-(3,4,5-trimethoxyphenyl)propanoic acid
Molecular Formula
C12H16O5
InChI Key
ZCYXGVJUZBKJAI-UHFFFAOYSA-N
SMILES
COC1=CC(CCC(O)=O)=CC(OC)=C1OC
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
100°C to 105°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text