Trimethylaluminium, 1.0M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Trimethylaluminium, 1.0M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimethylaluminium, 1.0M solution in heptane, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 189271000
Price (USD)/ Each
61.65
Online exclusive
71.80 
Save 10.15 (14%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)/ Each
61.65
Online exclusive
71.80 
Save 10.15 (14%)
Add to cart
Trimethylaluminium, 1.0M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Catalog number189271000
Price (USD)/ Each
61.65
Online exclusive
71.80 
Save 10.15 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Nametrimethylalumane
Molecular FormulaC3H9Al
InChI KeyJLTRXTDYQLMHGR-UHFFFAOYSA-N
SMILESC[Al](C)C
Molecular Weight (g/mol)72.09
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Titration Complexometric9.5 to 10.5 % w/w (on Al basis)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only