Tetrahydro-4H-pyran-4-one, 99%, Thermo Scientific Chemicals
Tetrahydro-4H-pyran-4-one, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrahydro-4H-pyran-4-one, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 189440250
Price (USD)/ Each
351.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
414.00 
Save 62.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
351.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
414.00 
Save 62.35 (15%)
Add to cart
Tetrahydro-4H-pyran-4-one, 99%, Thermo Scientific Chemicals
Catalog number189440250
Price (USD)/ Each
351.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
414.00 
Save 62.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameoxan-4-one
Molecular FormulaC5H8O2
InChI KeyJMJRYTGVHCAYCT-UHFFFAOYSA-N
SMILESO=C1CCOCC1
Molecular Weight (g/mol)100.12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4510 to 1.4530 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only