Chemical Identifiers
CAS16423-68-0
IUPAC Namedisodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular FormulaC20H6I4Na2O5
InChI KeyIINNWAYUJNWZRM-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12
View more